missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenium(IV) chloride, 99.5% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01309622

 View more versions of this product

Catalog No. AA1309622



Description

Description

It is used in the synthesis of other selenium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Selenium(IV) chloride
10026-03-6
100 g
Cl4Se
UN2923
selenium tetrachloride, selenium chloride secl4 , t-4, unii-4gb8868p5j, selenium iv chloride 1:4, secl4, tetrachloro-, e4-selane, selenium iv chloride, selenium iv chloride-8 mesh, selenium iv chloride trace metals basis 25g
LNBXMNQCXXEHFT-UHFFFAOYSA-N
220.771
220.77
Pungent
Moisture sensitive
(sublimation)
Lump
(metals basis)
MFCD00011225
14,8438
Decomposes in water and moist air; Insoluble in liquid bromine; Sublimes when heated
Cl[Se](Cl)(Cl)Cl
66205
99.5%
1.807
2.600 g/mL

Chemical Identifiers

10026-03-6
220.771
LNBXMNQCXXEHFT-UHFFFAOYSA-N
66205
Cl4Se
MFCD00011225
selenium tetrachloride, selenium chloride secl4 , t-4, unii-4gb8868p5j, selenium iv chloride 1:4, secl4, tetrachloro-, e4-selane, selenium iv chloride, selenium iv chloride-8 mesh, selenium iv chloride trace metals basis 25g
Cl[Se](Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 233-053-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only