Learn More
Selenium(IV) bromide, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04024006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Selenium(IV) bromide | |
7789-65-3 | |
5 g | |
MFCD00049766 | |
14,8437 | |
Decomposes in water; Soluble in carbon disulfide, chloroform, ethyl bromide | |
[Se](Br)(Br)(Br)Br | |
82246 | |
≥99% |
70°C to 80°C (decomposition) | |
Powder | |
Br4Se | |
UN2923 | |
selenium tetrabromide, selenium iv bromide, selenium bromide sebr4 , t-4, unii-8o54yxr180, tetrabromo-, e4-selane, sebr4, tetrabromo-$l^ 4-selane, tetrabromo-??-selane, selenium iv bromide 5g | |
VTQZBGAODFEJOW-UHFFFAOYSA-N | |
398.587 | |
398.6 | |
Moisture sensitive |
Chemical Identifiers
7789-65-3 | |
398.587 | |
VTQZBGAODFEJOW-UHFFFAOYSA-N | |
82246 |
Br4Se | |
MFCD00049766 | |
selenium tetrabromide, selenium iv bromide, selenium bromide sebr4 , t-4, unii-8o54yxr180, tetrabromo-, e4-selane, sebr4, tetrabromo-$l^ 4-selane, tetrabromo-??-selane, selenium iv bromide 5g | |
[Se](Br)(Br)(Br)Br |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber : 232-181-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only