missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenium ICP Standard, 1mL = 10mg Se (10,000ppm Se), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPSE10KN50

 View more versions of this product

Catalog No. PSE10KN-50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Selenium ICP Standard
86.0,11.57,0.86
Se
[Se]
78.971
CHEBI:27568
Poly natural
1 mL = 10 mg Se
50mL
7782-49-2,7697-37-2,7732-18-5
89.0,12.05,0.89
BUGBHKTXTAQXES-UHFFFAOYSA-N
selenium
6326970
Liquid
10,000ppm Se
Colorless

Chemical Identifiers

7782-49-2
78.971
6326970
selenium
Se
BUGBHKTXTAQXES-UHFFFAOYSA-N
CHEBI:27568
[Se]
Documents