Learn More
Selenium, 99+%, powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 419271000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
685°C | |
217°C | |
7782-49-2 | |
Powder | |
99% min | |
Se | |
01,990; 04,422; 05,575; 06,507; 08,439; 10,345; 11,465; 14,281 | |
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
selenium | |
6326970 | |
78.96 | |
Glass bottle | |
4.81 |
Selenium | |
<325 mesh | |
Cream-White to White | |
100 g | |
Se | |
MFCD00134090 MFCD00011224 | |
15, 8568 | |
Solubility in water: insoluble. Other solubilities: soluble in concentrated acid,slightly soluble in diehtyl ether,almost insoluble in carbon disulfide,insoluble in ethanol | |
[Se] | |
78.97 | |
CHEBI:27568 | |
≥99% | |
0.5% max | |
4.8100g/mL |
Chemical Identifiers
7782-49-2 | |
78.97 | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
6326970 | |
selenium |
Se | |
MFCD00134090 MFCD00011224 | |
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray | |
CHEBI:27568 | |
[Se] |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
May cause long lasting harmful effects to aquatic life.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 231-957-4
RUO – Research Use Only