Please login to your online account to display your discounted pricing

Selenium, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$117.02 - $117.02

Chemical Identifiers

Chemical Name or Material Selenium, 1000μg/mL (0.10% w/v)
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Se
Formula Weight 78.96
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003887

 
avantor performance
646504
Narrow Mouth Poly Bottle 150mL Each for $117.02
Description & Specifications

Specifications

Formula Weight 78.96
Chemical Name or Material Selenium, 1000μg/mL (0.10% w/v)
Molecular Formula Se
Packaging Narrow Mouth Poly Bottle
Quantity 150mL
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Specific Gravity 1L = 1.00kg