missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Selenic acid, 40% aq. soln.

Manufacturer:  Alfa Aesar™ 1885136

 View more versions of this product

Catalog No. AA1885136



Specifications

Specifications

Selenic acid
H10N2O4S
MFCD00012191
14,8429
[NH4+].[NH4+].O.[O-]S(=O)[O-]
134.15
144.97 (Anhydrous)
40% aq. soln.
Colorless
7783-11-1
H2SeO4
UN2922
ASPXBYAQZVXSNS-UHFFFAOYSA-N
diazanium;sulfite;hydrate
21701053
Liquid
1.41g/mL
500g

Chemical Identifiers

7783-11-1
134.15
ASPXBYAQZVXSNS-UHFFFAOYSA-N
diazanium;sulfite;hydrate
H10N2O4S
MFCD00012191
21701053
[NH4+].[NH4+].O.[O-]S(=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

TSCA : TSCA

SDS Documents