Selecto™ Preparative and Classic Column Chromatography Silica DFS Item
60Å gels made from identical starting material, making scale-up easier
$886.41 - $6,490.26
Specifications
Particle Size 63 to 200 μm
pH 7
Material Silica Gel
View More Specs
Products 2
Catalog Number Mfr. No. Particle Size Quantity Price Quantity  
Catalog Number Mfr. No. Particle Size Quantity Price Quantity  
05-719-822 DFS Item
View Documents Promotion Details
Selecto™
136644
63 to 200 μm 2.5 kg
Each for $886.41
05-719-823 DFS Item
View Documents Promotion Details
Selecto™
176644
63 to 200 μm 25 kg
Each for $6,490.26
Description
Description
  • pH of 7
  • Surface area of 500 to 600m2/g
Specifications
Specifications
63 to 200 μm
7
Silica Gel