Selecto™ Preparative and Classic Column Chromatography Silica
60Å gels made from identical starting material, making scale-up easier
Manufacturer: Selecto™ 126724
Description
- pH of 7
- Surface area of 500 to 600m2/g
Specifications
100 to 200 µm | |
7 | |
1 kg | |
Silica Gel |