Selecto™ Preparative and Classic Column Chromatography Silica
60Å gels made from identical starting material, making scale-up easier
Manufacturer: Selecto™ 136644
Description
- pH of 7
- Surface area of 500 to 600m2/g
Specifications
63 to 200 µm | |
7 | |
2.5 kg | |
Silica Gel |