Selecto™ Preparative and Classic Column Chromatography Silica DFS Item
60Å gels made from identical starting material, making scale-up easier
Manufacturer:  Selecto™ 126644
 View more versions of this product
Catalog No. 05-719-820
Description
Description
  • pH of 7
  • Surface area of 500 to 600m2/g
Specifications
Specifications
63 to 200 µm
7
500 g
Silica Gel