Selecto™ Preparative and Classic Column Chromatography Active Silica
60Å gels made from identical starting material, making scale-up easier
Supplier: Selecto™ 152824
Description
- pH of 7
- Surface area of 500 to 600m2/g
- Meets needs for standardized initial conditions and high reproducibility
- With 32-63μm particle size is ideal for fast, simple purifying of organic samples
Specifications
32 to 63 μm | |
7 | |
1 kg | |
Silica Gel |