Selecto™ Flexible-Backed TLC Plates DFS Item
Select from four media to match your separation needs
Manufacturer:  Selecto™ 31028
 View more versions of this product
Catalog No. 05-719-857
Description
Description
Select from four media to match your separation needs
  • Polymeric (cellulose, cellulose PEI, and alumina) and “G” (silica plates) binders are stable in aqueous solvents and virtually all mobile phases, and withstand many aggressive detection reagents
  • Flexible plastic backing for high durability—can be cut to any size required and stapled into a lab notebook; backing also makes it easy to isolate one spot for subsequent isolation/detection
  • Silica-gel plates separate weakly to strongly polar substances by adsorption chromatography
  • Cellulose plates for partition chromatography
  • Cellulose PEI plates separate weak anion exchangers such as amino acids and peptides (Store PEI plates at 4°C [39°F]; if stored at room temperature, plates may turn slightly yellow, affecting separation. Predevelop in distilled water to minimize yellow color.)
  • Alumina plates separate alkaloids, antibiotics, and hormones
Specifications
Specifications
50/Pk.
5 x 20 cm
Silica Gel
Widely Used for Dyes, Heterocyclic's, Pesticides, Herbicides, Fatty Acids, Amino Acids, Various Alcohols, Steroids, Hydrocarbons and Organic Synthesis Compounds
Silica Gel, Fluorescent Indicator
Plastic
200 µm