Selecto™ Flexible-Backed TLC Plates
Select from four media to match your separation needs
Manufacturer: Selecto™ 33328
Description
Select from four media to match your separation needs
- Polymeric (cellulose, cellulose PEI, and alumina) and “G” (silica plates) binders are stable in aqueous solvents and virtually all mobile phases, and withstand many aggressive detection reagents
- Flexible plastic backing for high durability—can be cut to any size required and stapled into a lab notebook; backing also makes it easy to isolate one spot for subsequent isolation/detection
- Silica-gel plates separate weakly to strongly polar substances by adsorption chromatography
- Cellulose plates for partition chromatography
- Cellulose PEI plates separate weak anion exchangers such as amino acids and peptides (Store PEI plates at 4°C [39°F]; if stored at room temperature, plates may turn slightly yellow, affecting separation. Predevelop in distilled water to minimize yellow color.)
- Alumina plates separate alkaloids, antibiotics, and hormones
Specifications
200/Pk. | |
2.5 x 7.5 cm | |
Silica Gel | |
Widely Used for Dyes, Heterocyclic's, Pesticides, Herbicides, Fatty Acids, Amino Acids, Various Alcohols, Steroids, Hydrocarbons and Organic Synthesis Compounds |
Silica Gel, Fluorescent Indicator | |
Plastic | |
200 µm |