Selecto™ Flexible-Backed TLC Plates DFS Item
Select from four media to match your separation needs
$825.47 - $879.53
Specifications
Backing Material Plastic
View More Specs
Products 3
Catalog Number Mfr. No. Type Quantity Price Quantity  
Catalog Number Mfr. No. Type Quantity Price Quantity  
05-719-859 DFS Item
View Documents Promotion Details
Selecto™
11089
Cellulose 300, Fluorescent Indicator 25/Pk.
Pack of 25 Each for $825.47
05-719-860 DFS Item
View Documents Promotion Details
Selecto™
10089
Cellulose 300, No Fluorescent Indicator 25/Pk.
Pack of 25 Each for $854.53
05-719-861 DFS Item
View Documents Promotion Details
Selecto™
10045
Alumina Basic, No Fluorescent Indicator 25/Pk.
Pack of 25 Each for $879.53
Description
Description
Select from four media to match your separation needs
  • Polymeric (cellulose, cellulose PEI, and alumina) and “G” (silica plates) binders are stable in aqueous solvents and virtually all mobile phases, and withstand many aggressive detection reagents
  • Flexible plastic backing for high durability—can be cut to any size required and stapled into a lab notebook; backing also makes it easy to isolate one spot for subsequent isolation/detection
  • Silica-gel plates separate weakly to strongly polar substances by adsorption chromatography
  • Cellulose plates for partition chromatography
  • Cellulose PEI plates separate weak anion exchangers such as amino acids and peptides (Store PEI plates at 4°C [39°F]; if stored at room temperature, plates may turn slightly yellow, affecting separation. Predevelop in distilled water to minimize yellow color.)
  • Alumina plates separate alkaloids, antibiotics, and hormones
Specifications
Specifications
Plastic