Sekisui Diagnostics OSOM™ Influenza A and B Test
Provides rapid differential diagnosis of Influenza A and B
$130.80 - $981.00
Specifications
For Use With (Application) Influenza A and B Testing
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
22071089 Sekisui Diagnostics Llc
SEK190
Influenza A and B Test 25 Tests Pack of 25 Each for $981.00
Case of 18 Pack for $17,658.00
22071090 Sekisui Diagnostics Llc
SEK191
Influenza A and B Control Kit Each Pack of 10 Each for $130.80
Specifications
Description
  • CLIA Complexity: Non-waived
    • CPT Code: 87804
    • Easy to read — three distinct lines
    • Nasal swab specimens
    • Two extra test sticks included for external QC testing
    • Less than one minute hands-on time, results in 10 minutes
    • Room temperature storage
    • Capable of detecting some animal and avian influenza strains
Description & Specifications
Specifications
Influenza A and B Testing