Labplas Secure-T Sample Bags
Made to handle the most robust blending applications
Manufacturer:  Labplas SCL4060FS
 View more versions of this product
Catalog No. 01-002-54
Description
Description
  • Durable
  • Can with stand blender use
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
  • Open top
Specifications
Specifications
Tear-off protection strip
3 mil
PE (Polyethylene)
152 x 102mm (6 x 4 in.)
205mL
Documents