missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Secure Click™ Nuisance Level Cartridges and Filters
Secure Click™ nuisance level cartridges and filters are NIOSH-approved for certain oil and non-oil based airborne particulate contaminants when used with 3M™ Secure Click™ facepieces.
Supplier: 3M™ 7100213359
Description
- Secure Click™ connection enables the quick and simple connection of filters
- Nuisance levels refer to concentrations not exceeding OSHA PEL, or other applicable government exposure limits, whichever is lower
- Dual-flow cartridges allow for easier breathing and greater comfort
- Swept-back design provides enhanced field of view and comfort
- For workplace and occupational applications only
Specifications
NIOSH-approved | |
Magenta | |
Mold Remediation | |
50 Filter Pairs | |
120 x 105 x 25 mm |
Secure Click™ | |
P100 | |
3M™ Secure Click™ Half Facepiece Reusable Respirators HF-800 Series | |
D3091 | |
15 g |