Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Secretory Component Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Secretory Component Ab-2, Mouse Monoclonal Antibody.

$238.04 - $667.27

Specifications

Antigen Secretory Component Ab-2
Clone COMPO2; same as SC-05
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1367P

 
thermo scientific™ lab vision™
MS1367P
1mL Each for $667.27

MS1367P0

 
thermo scientific™ lab vision™
MS1367P0
100μL Each for $238.04

MS1367P1

 
thermo scientific™ lab vision™
MS1367P1
500μL Each for $469.27

MS1367R7

 
thermo scientific™ lab vision™
MS1367R7
7mL Each for $469.27
Description & Specifications

Specifications

Antigen Secretory Component Ab-2
Clone COMPO2; same as SC-05
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Rat
Host Species Murine
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Semi- purified human secretory component (SC)
Isotype IgG1

Provide accurate, reproducible results with the Thermo Scientific™ Secretory Component Ab-2, Mouse Monoclonal Antibody.

Secretory Component is an epithelial transport receptor for polymeric IgA and IgM. Antibody to secretory component has been shown to be useful for identifying glandular carcinomas.

Host Species: Mouse

Clone: COMPO2; same as SC-05

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Semi-purified human secretory component (SC)

Molecular Weight: 80kDa

Positive Control: Small intestine, Colon CA, Stomach

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO