Please login to your online account to display your discounted pricing

sec-Butylbenzene, 99+%, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  107862500

 View more versions of this product

Catalog No. AC107862500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥99.0%
Formula Weight 134.22
Quantity 250mL
Color Clear
Beilstein 05, 414
Merck Index 15, 1552
EINECS Number 205-227-0
TSCA TSCA
RTECS Number CY9100000
Chemical Name or Material sec-Butylbenzene
Name Note 99+%
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C10H14
Refractive Index 1.4890 to 1.4910 (20°C, 589nm)
Linear Formula C6H5CH(CH3)C2H5
MDL Number MFCD00009329
CAS 135-98-8
Synonym 2-Phenylbutane
Water 0.05% max. (K.F.)
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Benzene 0.1% max. (GC)

Specifications

Chemical Name or Material sec-Butylbenzene
Name Note 99+%
CAS 135-98-8
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H14
Linear Formula C6H5CH(CH3)C2H5
Formula Weight 134.22
MDL Number MFCD00009329
Synonym 2-Phenylbutane
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents