missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

sec-Butylbenzene, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 107860500

 View more versions of this product

Catalog No. AC107860500


 

Specifications Description & Specifications

Specifications

Liquid
≥99.0%
Clear
15, 1552
2-Phenylbutane
99+%
C6H5CH(CH3)C2H5
sec-Butylbenzene
C10H14
0.05% max. (K.F.)
134.22
50mL
05, 414
0.1% max. (GC)
Glass bottle
MFCD00009329
99+%
135-98-8
1.4890 to 1.4910 (20°C, 589nm)
Authentic

Chemical Identifiers

135-98-8
C10H14
134.22
2-Phenylbutane
≥99.0%
99+%
C6H5CH(CH3)C2H5
MFCD00009329
sec-Butylbenzene, 99+%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-227-0

RTECSNumber : CY9100000

TSCA : TSCA