missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

sec-Butylamine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC107811000

 View more versions of this product

Catalog No. AC107811000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

sec-Butylamine
0.7200g/mL
Liquid
99%
C4H11N
C2H5CH(NH2)CH3
MFCD00008094
Clear colorless to light yellow
13952-84-6
Authentic
≥98.5%
Glass bottle
1.3920 to 1.3960 (20°C, 589nm)
73.13
2-Aminobutane
100mL

Chemical Identifiers

99%
C2H5CH(NH2)CH3
MFCD00008094
sec-Butylamine
C4H11N
73.13
2-Aminobutane
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger