Please login to your online account to display your discounted pricing

Alfa Aesar™ sec-Butyl methyl sulfide, 98%

Chemical Identifiers

Chemical Name or Material sec-Butyl methyl sulfide
CAS 10359-64-5
Molecular Formula C5H12S
Formula Weight 104.22
MDL Number MFCD00015193
UN Number UN1993
Beilstein 1730946
Synonym Methyl sec-butyl sulfide; 2-(Methylthio)butane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2194206

 
alfa aesar™
B2194206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2194214

 
alfa aesar™
B2194214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 104.22
Health Hazard 3 GHS P Statement
P210-P261-P243-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
Store in a well-ventilated place.
Chemical Name or Material sec-Butyl methyl sulfide
Assay 98%
Molecular Formula C5H12S
MDL Number MFCD00015193
CAS 10359-64-5
UN Number UN1993
Beilstein 1730946
Synonym Methyl sec-butyl sulfide; 2-(Methylthio)butane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Boiling Point 116° to 117°C
Quantity 5g
Flash Point 5°C (41°F)
Odor Unpleasant
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P261-P243-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
Store in a well-ventilated place.

GHS H Statement
H225
Highly flammable liquid and vapour.