missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ sec-Butyl methyl ether, 99%

Manufacturer:  Alfa Aesar™ H53427AC

 View more versions of this product

Catalog No. AAH53427AC



Description

Description

Applications
sec-Butyl methyl ether is used as a solvent in organic synthesis.

Solubility
Miscible with water, alcohol, ether and acetone.

Notes
Incompatible with strong oxidizing agents. Avoid conditions like flames, sparks and direct sunlight.
Specifications

Specifications

sec-Butyl methyl ether
57°C
6795-87-5
MFCD00190205
sec-butyl methyl ether, butane, 2-methoxy, methyl sec-butyl ether, ether, sec-butyl methyl, 2-butyl methyl ether, methyl 1-methylpropyl ether, s-butyl methyl ether, butane, 2-methoxy-9ci, butane,2-methoxy, acmc-1b64a
CCC(C)OC
88.15
88.15
−30°C (−22°F)
25mL
C5H12O
UN2350
FVNIMHIOIXPIQT-UHFFFAOYSA-N
2-methoxybutane
23238
99%

Chemical Identifiers

6795-87-5
88.15
FVNIMHIOIXPIQT-UHFFFAOYSA-N
23238
CCC(C)OC
C5H12O
MFCD00190205
sec-butyl methyl ether, butane, 2-methoxy, methyl sec-butyl ether, ether, sec-butyl methyl, 2-butyl methyl ether, methyl 1-methylpropyl ether, s-butyl methyl ether, butane, 2-methoxy-9ci, butane,2-methoxy, acmc-1b64a
2-methoxybutane
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL METHYL ETHER

RTECSNumber : KN5249500

SDS Documents