Learn More
sec-Butyl methyl ether, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H53427AE
Description
sec-Butyl methyl ether is used as a solvent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationssec-Butyl methyl ether is used as a solvent in organic synthesis.
Solubility
Miscible with water, alcohol, ether and acetone.
Notes
Incompatible with strong oxidizing agents. Avoid conditions like flames, sparks and direct sunlight.
Specifications
sec-Butyl methyl ether | |
0.742 | |
−30°C (−22°F) | |
1.372 | |
100 mL | |
sec-butyl methyl ether, butane, 2-methoxy, methyl sec-butyl ether, ether, sec-butyl methyl, 2-butyl methyl ether, methyl 1-methylpropyl ether, s-butyl methyl ether, butane, 2-methoxy-9ci, butane,2-methoxy, acmc-1b64a | |
FVNIMHIOIXPIQT-UHFFFAOYNA-N | |
2-methoxybutane | |
23238 | |
99% |
6795-87-5 | |
57°C | |
C5H12O | |
MFCD00190205 | |
UN2350 | |
Miscible with water,alcohol,ether and acetone. | |
CCC(C)OC | |
88.15 | |
88.15 |
Chemical Identifiers
6795-87-5 | |
88.15 | |
FVNIMHIOIXPIQT-UHFFFAOYNA-N | |
23238 | |
CCC(C)OC |
C5H12O | |
MFCD00190205 | |
sec-butyl methyl ether, butane, 2-methoxy, methyl sec-butyl ether, ether, sec-butyl methyl, 2-butyl methyl ether, methyl 1-methylpropyl ether, s-butyl methyl ether, butane, 2-methoxy-9ci, butane,2-methoxy, acmc-1b64a | |
2-methoxybutane |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL METHYL ETHER
RTECSNumber : KN5249500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only