missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

sec-Butyl acetate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1389222

 View more versions of this product

Catalog No. AAL1389222



Description

Description

sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications

Specifications

sec-Butyl acetate
-99°C
111°C to 112°C
C6H12O2
MFCD00009328
UN1123
14,1536
Slightly soluble in water.
CCC(C)OC(C)=O
116.16
116.16
105-46-4
0.872
16°C (60°F)
1.389
100 g
1720689
sec-butyl acetate, dl-sec-butyl acetate, 2-butyl acetate, acetic acid, 1-methylpropyl ester, 1-methylpropyl acetate, sec-butyl ethanoate, 1-methylpropyl ethanoate, acetic acid, sec-butyl ester, 2-butanol acetate, sec-butyl alcohol acetate
DCKVNWZUADLDEH-UHFFFAOYNA-N
butan-2-yl acetate
7758
98%

Chemical Identifiers

105-46-4
116.16
DCKVNWZUADLDEH-UHFFFAOYNA-N
7758
CCC(C)OC(C)=O
C6H12O2
MFCD00009328
sec-butyl acetate, dl-sec-butyl acetate, 2-butyl acetate, acetic acid, 1-methylpropyl ester, 1-methylpropyl acetate, sec-butyl ethanoate, 1-methylpropyl ethanoate, acetic acid, sec-butyl ester, 2-butanol acetate, sec-butyl alcohol acetate
butan-2-yl acetate
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 203-300-1

RTECSNumber : AF7380000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only