Please login to your online account to display your discounted pricing

sec-Butanol (Certified), Fisher Chemical


Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  O16641

 View more versions of this product

Catalog No. O1664-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Purity Grade Certified Grade
Boiling Point 99°C
Melting Point −115°C
Formula Weight 74.12
Quantity 1L
DOT Information DOT Class 3, : Flammable Liquid
Molecular Formula C4H10O
Density 0.800g/cm³
Vapor Pressure 17mbar at 20°C
Vapor Density 2.6g/mol
Viscosity 4.2 mPa/s at 20°C
Alpha Vector Butanol
Physical Form Liquid
Color Colorless
Packaging Glass Bottle
Boiling Range 1.5°C max., 98.0° to 101.0°C
Identification Pass Test
MDL Number MFCD00004569
Synonym 2-Butanol
Health Hazard 3 Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:2
Flammability:2
Instability:0
Chemical Name or Material sec-Butanol
Cleanup Symbol Solvent
ChemAlert Storage Symbol Red
Health Hazard 2 WARNING!
Linear Formula CH3CHOHCH2CH3
CAS 78-92-2

Specifications

Chemical Name or Material sec-Butanol
CAS 78-92-2
Purity Grade Certified Grade
Assay ≥99%
Molecular Formula C4H10O
Linear Formula CH3CHOHCH2CH3
Formula Weight 74.12
MDL Number MFCD00004569
Synonym 2-Butanol
Accessories
 

Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:2
Flammability:2
Instability:0

WARNING!

 

SDS & Safety Documents