Please login to your online account to display your discounted pricing

sec-Butanol (Certified), Fisher Chemical

$243.46 - $608.08

Chemical Identifiers

Chemical Name or Material sec-Butanol
CAS 78-92-2
Purity Grade Certified Grade
Assay ≥99%
Molecular Formula C4H10O
Linear Formula CH3CHOHCH2CH3
Formula Weight 74.12
MDL Number MFCD00004569
Synonym 2-Butanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

O1664-1

  SDS
fisher scientific company
O16641
Glass Bottle 1L Each for $243.46
Case of 6 Each for $1,460.76

O1664-4

  SDS
fisher scientific company
O16644
Glass Bottle 4L Each for $608.08
Case of 4 Each for $2,432.32
Description & Specifications

Specifications

Assay ≥99%
Formula Weight 74.12
Boiling Point 99°C
Melting Point −115°C
Purity Grade Certified Grade
Quantity 1L
DOT Information DOT Class 3, : Flammable Liquid
Molecular Formula C4H10O
Density 0.800g/cm³
Physical Form Liquid
Color Colorless
Viscosity 4.2 mPa/s at 20°C
Vapor Pressure 17mbar at 20°C
Vapor Density 2.6g/mol
Alpha Vector Butanol
Packaging Glass Bottle
Boiling Range 1.5°C max., 98.0° to 101.0°C
Synonym 2-Butanol
MDL Number MFCD00004569
Identification Pass Test
Health Hazard 3 Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:2
Flammability:2
Instability:0
Health Hazard 2 WARNING!
ChemAlert Storage Symbol Red
Cleanup Symbol Solvent
Chemical Name or Material sec-Butanol
Linear Formula CH3CHOHCH2CH3
CAS 78-92-2

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:2
Flammability:2
Instability:0

WARNING!