Please login to your online account to display your discounted pricing

sec-Butanol, 99+%, for analysis, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC220290025

 View more versions of this product

Catalog No. AC220290025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99%
Physical Form Liquid
Formula Weight 74.12
Quantity 2.5L
Color Clear
Purity Grade For analysis
Density 0.8100g/mL
Merck Index 15, 1543
Beilstein 01, 371
Iron (Fe) 0.5ppm max.
Synonym 2-Butanol
Chemical Name or Material sec-Butanol
Linear Formula C2H5CH(OH)CH3
Molecular Formula C4H10O
Name Note For analysis
CAS 78-92-2
Assay Percent Range 99+%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00004569
Copper (Cu) 0.1ppm max.
Water 0.1% max.
2_Propanol 0.2% max. (GC)
Non_volatile Matter 0.001% max.
Lead (Pb) 0.1ppm max.
Cadmium (Cd) 0.1ppm max.
Packaging Glass bottle
Free Acid 0.003% max.
Additional Information dibutyl ether (GC): < 0.2%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.
May cause respiratory irritation.

Specifications

Chemical Name or Material sec-Butanol
Name Note For analysis
CAS 78-92-2
Purity Grade For analysis
Assay ≥99%
Assay Percent Range 99+%
Molecular Formula C4H10O
Linear Formula C2H5CH(OH)CH3
Formula Weight 74.12
MDL Number MFCD00004569
Synonym 2-Butanol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.
May cause respiratory irritation.

SDS & Safety Documents