Please login to your online account to display your discounted pricing

Sebaconitrile 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  419251000

 View more versions of this product

Catalog No. AC419251000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 164.25
Physical Form Liquid
Quantity 100g
Color Clear colorless to light yellow
TSCA TSCA
RTECS Number VS1500000
EINECS Number 217-494-0
Density 0.91g/mL
Refractive Index 1.4470 to 1.4490 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Sebaconitrile
Name Note 98%
Molecular Formula C10H16N2
CAS 1871-96-1
Assay Percent Range 98%
Linear Formula NC(CH2)8CN
MDL Number MFCD00019900

Specifications

Chemical Name or Material Sebaconitrile
Name Note 98%
CAS 1871-96-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H16N2
Linear Formula NC(CH2)8CN
Formula Weight 164.25
MDL Number MFCD00019900
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents