MilliporeSigma™ Supelco™ SeaSpray™ Nebulizer, 1 mL/min. with EzyFit EZT-050 & EzyLok, Gas Flow 1 L/min.
Includes EZT-050 EzyFit Connector with 0.50 mm ID x 1.3 mm OD x 70 0 mm L tubing (1 unit) and EL-1 EzyLok Connector Kit
Supplier:  MilliporeSigma™ Supelco™ GXAR351FSS1E1EA
Catalog No. 111006637
Description
Description
  • This nebulizer supersedes AR35-1-FSS1
  • Suggested Gas Flow 1 L/min.
  • Suggested Sample Uptake 1 mL/min.
Specifications
Specifications
Nu Instruments Nu Plasma
Spectrometry
SDS
Documents
Documents
Promotions
Promotions
SeaSpray is a trademark of Glass Expansion Pty Ltd