missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sea sand, Washed, Thermo Scientific Chemicals
$68.69 - $68.69
Chemical Identifiers
CAS | 14808-60-7 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | silanedione |
SMILES | O=[Si]=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC612355000
|
Thermo Scientific Chemicals
612355000 |
500 g | Plastic bottle |
Each for $68.69
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidChemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
silanedione |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
14808-60-7 | |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
silanedione | |
24261 | |
60.08 | |
Plastic bottle |
500 g | |
SiO2 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Pass Test | |
Sand, White quartz |
RUO – Research Use Only