missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sea Sand, Extra Pure, MilliporeSigma™,

Manufacturer:  Emd Millipore Corporation 1077111000

Catalog No. M1077111000



Description

Description

Specifications

Specifications

14808-60-7
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
24261
60.08
Plastic Bottle
1713°C
1kg
O2Si
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite
O=[Si]=O
60.08
CHEBI:30563
2.65 g/3 (20 °C) not applicable
>2200°C (1013hPa)
5 to 8 (400g/L, H2O, 20°C) (slurry)

Chemical Identifiers

14808-60-7
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
silanedione
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite
CHEBI:30563
O=[Si]=O
Safety and Handling

Safety and Handling

EINECSNumber : 238-878-4

RTECSNumber : VV7330000

SDS
Documents