missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SDZ SER 082 fumarate, Tocris

Selective 5-HT2B/2C antagonist

Manufacturer:  R & D Systems 125550

 View more versions of this product

Catalog No. 125550


 

Specifications Description & Specifications

Specifications

SDZ SER 082 fumarate
>98%
344.41
50mg
1417343-80-6
C15H20N2·C4H4O4
(+)-cis-4,5,7a,8,9,10,11,11a-Octahydro-7H-10-methylindolo[1,7-bc][2,6]-naphthyridine fumarate

Chemical Identifiers

>98%
344.41
SDZ SER 082 fumarate
C15H20N2·C4H4O4
(+)-cis-4,5,7a,8,9,10,11,11a-Octahydro-7H-10-methylindolo[1,7-bc][2,6]-naphthyridine fumarate

Safety and Handling

Recommended Storage : Store at Room Temperature