Please login to your online account to display your discounted pricing

SDZ SER 082 fumarate, Tocris


Selective 5-HT2B/2C antagonist

Quantity 10mg

Manufacturer: r & d systems  125510

 View more versions of this product

Catalog No. 125510

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 344.41
Quantity 10mg
Assay Percent Range >98%
Chemical Name or Material SDZ SER 082 fumarate
CAS 1417343-80-6
Molecular Formula C15H20N2·C4H4O4
Synonym (+)-cis-4,5,7a,8,9,10,11,11a-Octahydro-7H-10-methylindolo[1,7-bc][2,6]-naphthyridine fumarate
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material SDZ SER 082 fumarate
Assay Percent Range >98%
Molecular Formula C15H20N2·C4H4O4
Formula Weight 344.41
Synonym (+)-cis-4,5,7a,8,9,10,11,11a-Octahydro-7H-10-methylindolo[1,7-bc][2,6]-naphthyridine fumarate
Accessories

Store at Room Temperature