Please login to your online account to display your discounted pricing

SDZ SER 082 fumarate, Tocris

Selective 5-HT2B/2C antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material SDZ SER 082 fumarate
Assay Percent Range >98%
Molecular Formula C15H20N2·C4H4O4
Formula Weight 344.41
Synonym (+)-cis-4,5,7a,8,9,10,11,11a-Octahydro-7H-10-methylindolo[1,7-bc][2,6]-naphthyridine fumarate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

125510

 
r & d systems
125510
10mg Each for $239.63

125550

 
r & d systems
125550
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 344.41
Quantity 10mg
Chemical Name or Material SDZ SER 082 fumarate
Assay Percent Range >98%
Molecular Formula C15H20N2·C4H4O4
CAS 1417343-80-6
Synonym (+)-cis-4,5,7a,8,9,10,11,11a-Octahydro-7H-10-methylindolo[1,7-bc][2,6]-naphthyridine fumarate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature