Please login to your online account to display your discounted pricing

SDZ 220-581, Tocris


Competitive NMDA antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS125010

 View more versions of this product

Catalog No. 125010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 369.74
Quantity 10mg
Molecular Formula C16H17ClNO5P
Chemical Name or Material SDZ 220-581
Synonym (S)-α-Amino-2'-chloro-5-(phosphonomethyl)[1, 1'-biphenyl]-3-propanoic acid
CAS 174575-17-8
Assay Percent Range >98%
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material SDZ 220-581
Assay Percent Range >98%
Molecular Formula C16H17ClNO5P
Formula Weight 369.74
Synonym (S)-α-Amino-2'-chloro-5-(phosphonomethyl)[1, 1'-biphenyl]-3-propanoic acid
Accessories

Store at Room Temperature