Please login to your online account to display your discounted pricing

SDZ 220-581, Tocris

Competitive NMDA antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material SDZ 220-581
Assay Percent Range >98%
Molecular Formula C16H17ClNO5P
Formula Weight 369.74
Synonym (S)-α-Amino-2'-chloro-5-(phosphonomethyl)[1, 1'-biphenyl]-3-propanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

125010

 
r & d systems
RDS125010
10mg Each for $252.39

125050

 
r & d systems
RDS125050
50mg Each for $1,050.42
Description & Specifications

Specifications

Chemical Name or Material SDZ 220-581
CAS 174575-17-8
Recommended Storage Store at Room Temperature
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C16H17ClNO5P
Formula Weight 369.74
Synonym (S)-α-Amino-2'-chloro-5-(phosphonomethyl)[1, 1'-biphenyl]-3-propanoic acid
Safety and Handling

Store at Room Temperature