Please login to your online account to display your discounted pricing

SDZ 220-040, Tocris

Potent, competitive NMDA antagonist

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material SDZ 220-040
Assay Percent Range >98%
Molecular Formula C16H16Cl2NO6P
Formula Weight 420.19
Synonym (S)-α-Amino-2', 4'-dichloro-4-hydroxy-5-(phosphonomethyl)-[1, 1'-biphenyl]-3-propanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

125110

 
r & d systems
RDS125110
10mg Each for $257.85

125150

 
r & d systems
RDS125150
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 420.19
Quantity 10mg
Assay Percent Range >98%
Chemical Name or Material SDZ 220-040
Molecular Formula C16H16Cl2NO6P
CAS 174575-40-7
Synonym (S)-α-Amino-2', 4'-dichloro-4-hydroxy-5-(phosphonomethyl)-[1, 1'-biphenyl]-3-propanoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature