Please login to your online account to display your discounted pricing

SDZ 21009, Tocris

β-adrenoceptor antagonist. Also 5-HT1A/1B antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material SDZ 21009
Assay Percent Range >98%
Molecular Formula C19H28N2O4
Formula Weight 348.44
Synonym 4-[3-[(1, 1-Dimethylethyl)amino]-2-hydroxypropoxy]-1H-indole-2-carboxylic acid, 1-methylethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

151610

 
r & d systems
151610
10mg Each for $239.63

151650

 
r & d systems
151650
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 348.44
Quantity 10mg
Chemical Name or Material SDZ 21009
Assay Percent Range >98%
Molecular Formula C19H28N2O4
Synonym 4-[3-[(1, 1-Dimethylethyl)amino]-2-hydroxypropoxy]-1H-indole-2-carboxylic acid, 1-methylethyl ester
Recommended Storage Store at Room Temperature
CAS 39731-05-0
Safety and Handling

Store at Room Temperature