Please login to your online account to display your discounted pricing

SDZ 205-557 hydrochloride, Tocris

5-HT4/5-HT3 receptor antagonist

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material SDZ 205-557 hydrochloride
Assay Percent Range >99%
Molecular Formula C14H21ClN2O3·HCl
Formula Weight 337.25
Synonym 4-Amino-5-chloro-2-methoxybenzoic acid 2-(diethylamino)ethyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

203750

 
r & d systems
RDS203750
50mg Each for $697.40

203710

 
r & d systems
RDS203710
10mg Each for $168.15
Description & Specifications

Specifications

Formula Weight 337.25
Quantity 50mg
Chemical Name or Material SDZ 205-557 hydrochloride
Assay Percent Range >99%
Molecular Formula C14H21ClN2O3·HCl
CAS 1197334-02-3
Synonym 4-Amino-5-chloro-2-methoxybenzoic acid 2-(diethylamino)ethyl ester hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature