Please login to your online account to display your discounted pricing

SDM25N hydrochloride, Tocris

Potent, selective non-peptide δ antagonist

$214.12 - $891.94

Chemical Identifiers

Chemical Name or Material SDM25N hydrochloride
Assay Percent Range >99%
Molecular Formula C26H26N2O3·HCl
Formula Weight 450.96
Synonym (4bS,8R,8aS,14bR)-5,6,7,8,14,14b-Hexahydro-7-(2-methyl-2-propenyl)-4,8-methanobenzofuro[2,3-a]pyrido[4,3-b]carbazole-1,8a(9H)-diol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

141010

 
r & d systems
RDS141010
10mg Each for $214.12

141050

 
r & d systems
RDS141050
50mg Each for $891.94
Description & Specifications

Specifications

Formula Weight 450.96
Quantity 10mg
Assay Percent Range >99%
CAS 342884-71-3
Synonym (4bS,8R,8aS,14bR)-5,6,7,8,14,14b-Hexahydro-7-(2-methyl-2-propenyl)-4,8-methanobenzofuro[2,3-a]pyrido[4,3-b]carbazole-1,8a(9H)-diol hydrochloride
Chemical Name or Material SDM25N hydrochloride
Molecular Formula C26H26N2O3·HCl
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C