Please login to your online account to display your discounted pricing

SD 208, Tocris

Potent ATP-competitive TGF-βRI inhibitor

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material SD 208
Assay Percent Range >98%
Molecular Formula C17H10ClFN6
Formula Weight 352.75
Synonym 2-(5-Chloro-2-fluorophenyl)-4-[(4-pyridyl)amino]pteridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

326950

 
r & d systems
326950
50mg Each for $895.76

326910

 
r & d systems
326910
10mg Each for $214.12
Description & Specifications

Specifications

Formula Weight 352.75
Quantity 50mg
Chemical Name or Material SD 208
Assay Percent Range >98%
CAS 627536-09-8
Synonym 2-(5-Chloro-2-fluorophenyl)-4-[(4-pyridyl)amino]pteridine
Molecular Formula C17H10ClFN6
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature