missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

scyllo-Inositol, TCI America™

$100.83 - $300.98

Chemical Identifiers

CAS 488-59-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00065455
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0631200MG View Documents TCI America
I0631200MG
200mg Each for $100.83
 
I06311G View Documents TCI America
I06311G*
1g Each for $300.98
 
Specifications

Chemical Identifiers

488-59-5
180.156
CDAISMWEOUEBRE-UHFFFAOYSA-N
CHEBI:24848
C1(C(C(C(C(C1O)O)O)O)O)O
C6H12O6
MFCD00065455
892
cyclohexane-1,2,3,4,5,6-hexol

Specifications

White
200mg
C6H12O6
CDAISMWEOUEBRE-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexol
892
180.16
≥99.0% (HPLC)
350°C
488-59-5
MFCD00065455
C1(C(C(C(C(C1O)O)O)O)O)O
180.156
CHEBI:24848
Crystal-Powder at 20°C
scyllo-Inositol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3265

TSCA : No

Recommended Storage : Freezer