missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Scriptaid

Histone deacetylase inhibitor

$168.15 - $697.40

Specifications

Product Type Scriptaid
Inhibitors Scriptaid
Molecular Weight 326.35
Purity >99
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
242110 Tocris™
RDS242110
10mg Each for $168.15
242150 Tocris™
RDS242150
50mg Each for $697.40
Specifications

Description

Novel histone deacetylase inhibitor. Produces > 100-fold increase in histone acetylation and displays relatively low toxicity.
  • Chemical Name: N-Hydroxy-1,3-dioxo-1H-benz[de]isoquinoline-2(3H)-hexanamide
  • Formula: C18H18N2O4
  • CAS Number: 287383-59-9
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99
Description & Specifications

Specifications

Scriptaid
Scriptaid
326.35
>99