Please login to your online account to display your discounted pricing

Thermo Scientific™ Screw Cap Septum Vials


For a convenient, versatile, and cost-effective vial and closure system, choose Thermo Scientific™ Screw Cap Septum Vials.

Manufacturer: thermo scientific™  TS13043

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. PI13043

  • / Pack of 72 Each
 

Description & Specifications

Specifications

Diameter (Metric) Outer 21mm
Type Screw Cap Septum Vial
Volume (Metric) 14mL
Height (Metric) 70mm
Closure Size 18-400mm
Description 14mL; Clear

For a convenient, versatile, and cost-effective vial and closure system, choose Thermo Scientific™ Screw Cap Septum Vials. Made of autoclavable borosilicate glass and available with a wide assortment of special closures and accessories, these vials are ideal for sample collection and small derivatization reactions for use with automated gas (GC) and liquid chromatography (LC), and for storage of reagents and standards under complete seal with instant syringe access. The heavy-duty, flip-top divider box provides easy access to vials, caps, and septa and offers a convenient sample storage center. Clear and amber vials are available.

Applications

The 40mL clear or amber vial with a Thermo Scientific Disc is suitable for discrete water sampling under U.S. EPA 40 CFR Parts 136 and 141.

The 1.5mL clear and amber vials fit GC and LC autosamplers that use standard 12 ´ 32 mm vials.

Accessories

Accessories for Catalog Number: PI13043

Item Mfr. No. Item Description Price Per Unit Quantity