Please login to your online account to display your discounted pricing

ScintiVerse™ BD Cocktail (Scintanalyzed™), Fisher Chemical


Universal LSC Cocktail for aqueous, nonaqueous and emulsion counting systems. Contains biodegradable linear alkyl aromatic solvent, which can be classified as nonhazardous liquid by local sewerage regulations

Quantity 250mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  SX18250

 View more versions of this product

Catalog No. SX18-250

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Quantity 250mL
Physical Form Liquid
Water 0.7% Max.
Purity Grade Scintanalyzed™
Specific Gravity 0.890 to 0.910 (at 25°C)
CAS 92-71-7
LSC Efficiency 3H 0.60 min.
CAS 9016-45-9
Background LSC Analysis 50 CPM max.
Chemical Name or Material ScintiVerse™ BD Cocktail
CAS 67774-74-7
MDL Number MFCD00212667
Packaging Glass Bottle
LSC Analysis 0.55 min.
CAS 13280-61-0
CAS 577-11-7
Flash Point 279°F min.
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material ScintiVerse™ BD Cocktail
CAS 67774-74-7
CAS 577-11-7
CAS 9016-45-9
CAS 92-71-7
CAS 13280-61-0
Purity Grade Scintanalyzed™
MDL Number MFCD00212667
Accessories
 

SDS & Safety Documents