missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ScintiVerse™ BD Cocktail (Scintanalyzed™), Fisher Chemical

Universal LSC Cocktail for aqueous, nonaqueous and emulsion counting systems. Contains biodegradable linear alkyl aromatic solvent, which can be classified as nonhazardous liquid by local sewerage regulations

$163.71 - $780.93

Chemical Identifiers

CAS 67774-74-7, 577-11-7, 9016-45-9, 92-71-7,13280-61-0,577-11-7,9016-45-9,92-71-7,13280-61-0
MDL Number MFCD00212667
Chemical Name or Material ScintiVerse™ BD Cocktail
Grade Scintanalyzed™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
SX18-250 SDS Fisher Scientific Company
SX18250
Glass Bottle 250mL Each for $163.71
Case of 6 Each for $982.26
SX18-4 SDS Accessories Fisher Scientific Company
FLSX184
Glass Bottle 4L Each for $780.93
Case of 4 Each for $3,123.72
Specifications

Description

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Description & Specifications

Chemical Identifiers

67774-74-7, 577-11-7, 9016-45-9, 92-71-7,13280-61-0,577-11-7,9016-45-9,92-71-7,13280-61-0
MFCD00212667
ScintiVerse™ BD Cocktail
Scintanalyzed™

Specifications

250mL
50 CPM max.
0.55 min.
0.60 min.
MFCD00212667
0.7% Max.
Scintanalyzed™
67774-74-7,577-11-7,9016-45-9,92-71-7,13280-61-0
279°F min.
Liquid
Glass Bottle
0.890 to 0.910 (at 25°C)
ScintiVerse™ BD Cocktail

Safety and Handling