Please login to your online account to display your discounted pricing

Science Literacy Series


Explore essential concepts with this NSES-based series.

Manufacturer: fisher science education  #033157

 View more versions of this product

Catalog No. S66853

  • / Each
 

Description & Specifications

Specifications

Product Type Student Physics Science Book
Author Maxine Levaren
Category Physics
For Use With (Application) Students with low reading levels
Format Book
Grade 6 to 8