Please login to your online account to display your discounted pricing

Science Literacy Series


Explore essential concepts with this NSES-based series.

Manufacturer: fisher science education  #033114

 View more versions of this product

Catalog No. S66840

  • / Each
 

Description & Specifications

Specifications

Category Chemistry
Format Book
Grade 5 to 9
Product Type Student Chemistry Book
For Use With (Application) Students with low reading levels
Author David G. Bauer