Please login to your online account to display your discounted pricing

Science Literacy Series


Explore essential concepts with this NSES-based series.

Manufacturer: fisher science education  #033130

 View more versions of this product

Catalog No. S66838

  • / Each
 

Description & Specifications

Specifications

Author George Watson
Category Biology
For Use With (Application) Students with low reading levels
Format Book
Grade 5 to 9
Product Type Student Biology Book

  • Promote critical thinking and science process skills
  • Designed for all students with low reading levels
  • Teacher's edition includes background information, assessments, and answer keys
  • Student books: 1997-2000; 64 pages
  • Teacher's guides: 1997-2000; 96 pages
  • Grades 5-9

  • Biology—Human biology, genetic engineering, ecology, zoology, and botany
  • Chemistry—Atomic structure, properties of matter, acids and bases, solutions, and a variety of scientific processes
  • Earth Science—Rocks and minerals, continental drift, volcanoes, earthquakes, weather, and the water cycle
  • Physics—Electricity and magnetism, light, and energy